Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG2-alkyne | 2227450-68-0 | MFCD31922733 | 95.6% | 369.48 g/mol | C17H27N3O4S | 100 MG
SDP

Catalog No. 5000284247
Encompass_Preferred

Biotin-PEG2-alkyne is a short polyethylene glycol (PEG) linker functionalized with a biotin group and a terminal alkyne. It is designed for affinity-based capture and copper(I)-catalyzed azide-alkyne cycloaddition (click chemistry) conjugations in research applications.

  • Contains a biotin moiety for affinity capture.
  • Includes a terminal alkyne for copper(I)-catalyzed click chemistry.
  • Short PEG2 linker improves solubility and reduces steric hindrance.
  • Supplied as an off-white to light yellow solid.
  • Purity 95.6% by supplier specification.
  • Suitable for pull-down assays and biotinylation workflows.
  • Store powder at -20°C for long-term stability; 4°C for shorter storage.

Catalog No. 50-002-84247 Supplier Medchemexpress LLC Supplier No. HYW615095100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG2-alkyne is a short polyethylene glycol (PEG) linker functionalized with a biotin group and a terminal alkyne. It is designed for affinity-based capture and copper(I)-catalyzed azide-alkyne cycloaddition (click chemistry) conjugations in research applications.

  • Contains a biotin moiety for affinity capture.
  • Includes a terminal alkyne for copper(I)-catalyzed click chemistry.
  • Short PEG2 linker improves solubility and reduces steric hindrance.
  • Supplied as an off-white to light yellow solid.
  • Purity 95.6% by supplier specification.
  • Suitable for pull-down assays and biotinylation workflows.
  • Store powder at -20°C for long-term stability; 4°C for shorter storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.