Learn More
Medchemexpress LLC Biotin-PEG2-azide | 1910803-72-3 | 97.0% | 250 MG
SDP
Catalog No. 5000343021
Encompass_Preferred
Biotin-PEG2-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules containing DBCO or BCN groups. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
  • Useful in cancer research and targeted therapy
Catalog No. 50-003-43021 Supplier Medchemexpress LLC Supplier No. 250MGHY126957
Biotin-PEG2-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules containing DBCO or BCN groups. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
  • Useful in cancer research and targeted therapy