Learn More
TARGETMOL CHEMICALS INC Biotin-PEG2-C2-iodoacetamide
SDP
Catalog No. 502878366
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Biotin-PEG2-C2-iodoacetamide (Biotin-PEG2-iodoacetamide) is a PEG-based PROTAC linker that can be used in PROTAC synthesis. Purity 98.01%
Catalog No. 50-287-8366 Supplier TARGETMOL CHEMICALS INC Supplier No. T1756550MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Biotin-PEG2-C2-iodoacetamide (Biotin-PEG2-iodoacetamide) is a PEG-based PROTAC linker that can be used in PROTAC synthesis. Purity 98.01%