Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG2-C6-azide 100mg | 1011268-29-3 | 513.65 g/mol | C22H39N7O5S | 100 MG
SDP

Catalog No. 5000308188
Encompass_Preferred

Biotin-PEG2-C6-azide is a biotinylated polyethylene glycol linker bearing a terminal azide functional group used for bioconjugation, click chemistry, and PROTAC linker synthesis.

  • Provides a terminal azide for copper-catalyzed azide-alkyne cycloaddition.
  • Contains a PEG2 spacer to improve solubility and reduce steric hindrance.
  • Biotin tag enables streptavidin-based capture and detection workflows.
  • Molecular formula C22H39N7O5S; molecular weight ~513.65 g/mol.
  • CAS number 1011268-29-3.
  • Purity typically ≥98%; appearance white solid; store at -20 °C.
  • Available in small-scale research quantities (e.g., 2 mg, 10 mg, 100 mg), packaging varies by supplier.

Catalog No. 50-003-08188 Supplier Medchemexpress LLC Supplier No. HY117042100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG2-C6-azide is a biotinylated polyethylene glycol linker bearing a terminal azide functional group used for bioconjugation, click chemistry, and PROTAC linker synthesis.

  • Provides a terminal azide for copper-catalyzed azide-alkyne cycloaddition.
  • Contains a PEG2 spacer to improve solubility and reduce steric hindrance.
  • Biotin tag enables streptavidin-based capture and detection workflows.
  • Molecular formula C22H39N7O5S; molecular weight ~513.65 g/mol.
  • CAS number 1011268-29-3.
  • Purity typically ≥98%; appearance white solid; store at -20 °C.
  • Available in small-scale research quantities (e.g., 2 mg, 10 mg, 100 mg), packaging varies by supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.