Learn More
Medchemexpress LLC Biotin-PEG2-C6-azide 10mg | 1011268-29-3 | 513.7 g/mol | C22H39N7O5S | 10 MG
SDP
Catalog No. 5000229649
Encompass_Preferred
Biotin-PEG2-C6-azide is a biotinylated polyethylene glycol (PEG) linker with a C6 spacer and a terminal azide group. It is supplied in small quantities for research use and is used for click chemistry conjugation, affinity capture, and as a PROTAC linker in chemical biology and proteomics workflows.
  • Provides a biotin handle for affinity capture with streptavidin or avidin.
  • Contains a terminal azide for copper-catalyzed or strain-promoted azide-alkyne cycloaddition.
  • PEG2 spacer improves solubility and reduces steric hindrance.
  • C6 linker offers flexible spacing between biotin and the reactive group.
  • Supplied in small research quantities suitable for synthesis and labeling workflows.
Catalog No. 50-002-29649 Supplier Medchemexpress LLC Supplier No. HY11704210MG
Biotin-PEG2-C6-azide is a biotinylated polyethylene glycol (PEG) linker with a C6 spacer and a terminal azide group. It is supplied in small quantities for research use and is used for click chemistry conjugation, affinity capture, and as a PROTAC linker in chemical biology and proteomics workflows.
  • Provides a biotin handle for affinity capture with streptavidin or avidin.
  • Contains a terminal azide for copper-catalyzed or strain-promoted azide-alkyne cycloaddition.
  • PEG2 spacer improves solubility and reduces steric hindrance.
  • C6 linker offers flexible spacing between biotin and the reactive group.
  • Supplied in small research quantities suitable for synthesis and labeling workflows.