Learn More
Medchemexpress LLC Biotin-PEG2-Mal | 305372-39-8 | 99.6% | 525.62 | 100 MG
SDP
Catalog No. 5000453526
Encompass_Preferred
Biotin-PEG2-Mal is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which contain two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTAC.
Catalog No. 50-004-53526 Supplier Medchemexpress LLC Supplier No. HYW010764100MG
Biotin-PEG2-Mal is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which contain two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTAC.