Learn More
Medchemexpress LLC Biotin-PEG2-NH-Boc | 175885-18-4 | C21H38N4O6S | 1 G
SDP
Catalog No. 5000354894
Encompass_Preferred
Biotin-PEG2-NH-Boc is identified as a biotin-labeled, PEG-based linker used in the synthesis of PROTACs. These PROTACs are compounds designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Functions as a linker for targeted protein degradation research.
  • Utilizes a biotin label.
  • Peg-based structure.
  • Acts through the ubiquitin-proteasome system.
  • Facilitates selective protein degradation.
Catalog No. 50-003-54894 Supplier Medchemexpress LLC Supplier No. HY1409341G
Biotin-PEG2-NH-Boc is identified as a biotin-labeled, PEG-based linker used in the synthesis of PROTACs. These PROTACs are compounds designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Functions as a linker for targeted protein degradation research.
  • Utilizes a biotin label.
  • Peg-based structure.
  • Acts through the ubiquitin-proteasome system.
  • Facilitates selective protein degradation.