Learn More
Medchemexpress LLC Biotin-PEG2-NH-Boc | 175885-18-4 | C21H38N4O6S | 100 MG
Supplier: Medchemexpress LLC HY140934100MG
Biotin-PEG2-NH-Boc is a biotin-labeled, PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It is intended for research purposes only. PROTACs function by utilizing the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins, achieved through two different ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other the protein of interest.
- PROTAC linker
- Useful in cancer research and targeted therapy