Learn More
Medchemexpress LLC Biotin-PEG2-NH-Boc | 175885-18-4 | C21H38N4O6S | 250 MG
SDP
Catalog No. 5000352778
Encompass_Preferred
Biotin-PEG2-NH-Boc is a biotin-labeled, PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. These PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. It is intended strictly for research purposes.
  • Biotin-labeled, PEG-based linker
  • Used in PROTAC synthesis
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-52778 Supplier Medchemexpress LLC Supplier No. 250MGHY140934
Biotin-PEG2-NH-Boc is a biotin-labeled, PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. These PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. It is intended strictly for research purposes.
  • Biotin-labeled, PEG-based linker
  • Used in PROTAC synthesis
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only