Learn More
Medchemexpress LLC Biotin-PEG2-NH-Boc | 175885-18-4 | C21H38N4O6S | 50 MG
SDP
Catalog No. 5000354587
Encompass_Preferred
Biotin-PEG2-NH-Boc is a biotin-labeled, PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This linker is essential for constructing PROTAC molecules, which function by connecting an E3 ubiquitin ligase ligand to a target protein ligand, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • Biotin-labeled for specific applications
  • PEG-based linker for PROTAC synthesis
  • Utilizes the ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-54587 Supplier Medchemexpress LLC Supplier No. HY14093450MG
Biotin-PEG2-NH-Boc is a biotin-labeled, PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This linker is essential for constructing PROTAC molecules, which function by connecting an E3 ubiquitin ligase ligand to a target protein ligand, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • Biotin-labeled for specific applications
  • PEG-based linker for PROTAC synthesis
  • Utilizes the ubiquitin-proteasome system for selective protein degradation