Learn More
Medchemexpress LLC Biotin-PEG2-NHS ester | 596820-83-6 | 96.7% | C21H32N4O8S | 100 MG
SDP
Catalog No. 5000355571
Encompass_Preferred
Biotin-PEG2-NHS ester is a biotin-labeled, PEG-based PROTAC linker designed for research use. It can be utilized in the synthesis of PROTACs, which employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs consist of two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55571 Supplier Medchemexpress LLC Supplier No. HY140888100MG
Biotin-PEG2-NHS ester is a biotin-labeled, PEG-based PROTAC linker designed for research use. It can be utilized in the synthesis of PROTACs, which employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs consist of two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins