Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG2-NHS ester | 596820-83-6 | 96.7% | C21H32N4O8S | 50 MG
SDP

Catalog No. 5000355001
Encompass_Preferred

Biotin-PEG2-NHS ester is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs.

  • For research use only
  • White to off-white solid appearance
  • Molecular weight of 500.57
  • Powder can be stored at -20°C for 3 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 125 mg/mL

Catalog No. 50-003-55001 Supplier Medchemexpress LLC Supplier No. HY14088850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG2-NHS ester is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs.

  • For research use only
  • White to off-white solid appearance
  • Molecular weight of 500.57
  • Powder can be stored at -20°C for 3 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 125 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.