Learn More
Medchemexpress LLC Biotin-PEG3-acid 100mg | 252881-76-8 | 100 MG
SDP
Catalog No. 5000229773
Encompass_Preferred
Biotin-PEG3-acid is a biotin-labeled polyethylene glycol (PEG3) linker containing a terminal carboxylic acid. It is supplied as a purified research-grade small molecule for use in PROTAC synthesis and biotinylation applications.
  • Biotin-labeled PEG3 linker with terminal carboxylic acid.
  • Designed for PROTAC synthesis and biotinylation workflows.
  • High purity, 98.0%.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Store sealed at 4°C; in solution store at -80°C for long-term stability.
  • Available in 100 MG and larger pack sizes for research use.
Catalog No. 50-002-29773 Supplier Medchemexpress LLC Supplier No. HY116027100MG
Biotin-PEG3-acid is a biotin-labeled polyethylene glycol (PEG3) linker containing a terminal carboxylic acid. It is supplied as a purified research-grade small molecule for use in PROTAC synthesis and biotinylation applications.
  • Biotin-labeled PEG3 linker with terminal carboxylic acid.
  • Designed for PROTAC synthesis and biotinylation workflows.
  • High purity, 98.0%.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Store sealed at 4°C; in solution store at -80°C for long-term stability.
  • Available in 100 MG and larger pack sizes for research use.