Learn More
Medchemexpress LLC Biotin-PEG3-acid 10g | 252881-76-8 | 447.55 g·mol⁻¹ | C19H33N3O7S | 10 G
SDP
Supplier:  Medchemexpress LLC HY11602710G
Encompass_Preferred
Biotin-PEG3-acid is a biotin-labeled PEG-based linker with a terminal carboxylic acid, supplied as a solid for research use. It is designed for conjugation to amine-containing molecules via amide coupling and provides a biotin tag for affinity capture, detection, or purification workflows.
  • Biotinylated PEG3 spacer with terminal carboxylic acid for amide coupling.
  • Provides a short, flexible spacer to reduce steric hindrance in bioconjugates.
  • Enhances water solubility compared with biotin alone.
  • Suitable for PROTAC linker synthesis, biotinylation, affinity purification, and detection.
  • Supplied as a high-purity solid appropriate for research applications.
Catalog No. 50-002-27899