Learn More
Medchemexpress LLC Biotin-PEG3-acid 1g | 252881-76-8 | 447.55 | C19H33N3O7S | 1 G
SDP
Catalog No. 5000226384
Encompass_Preferred
Biotin-PEG3-acid is a biotin-labeled, PEG-based PROTAC linker used to introduce a biotin tag with a short PEG3 spacer in PROTAC synthesis and bioconjugation workflows. The material is supplied as a 1 g purified solid (typical purity 98.0%) with molecular weight 447.55 g/mol and chemical formula C19H33N3O7S, and is accompanied by a batch-specific certificate of analysis.
  • Provides a biotin handle separated by a PEG3 spacer to reduce steric hindrance
  • Designed for incorporation into PROTACs and other bioconjugates
  • Improves aqueous solubility compared with direct biotinylation
  • Typically supplied with batch certificate showing ~98.0% purity
  • Solid form with known molecular weight and formula for formulation planning
Catalog No. 50-002-26384 Supplier Medchemexpress LLC Supplier No. HY1160271G
Biotin-PEG3-acid is a biotin-labeled, PEG-based PROTAC linker used to introduce a biotin tag with a short PEG3 spacer in PROTAC synthesis and bioconjugation workflows. The material is supplied as a 1 g purified solid (typical purity 98.0%) with molecular weight 447.55 g/mol and chemical formula C19H33N3O7S, and is accompanied by a batch-specific certificate of analysis.
  • Provides a biotin handle separated by a PEG3 spacer to reduce steric hindrance
  • Designed for incorporation into PROTACs and other bioconjugates
  • Improves aqueous solubility compared with direct biotinylation
  • Typically supplied with batch certificate showing ~98.0% purity
  • Solid form with known molecular weight and formula for formulation planning