Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG3-acid 250mg | 252881-76-8 | 447.55 g·mol⁻¹ | C19H33N3O7S | 250 MG
SDP

Catalog No. 5000229481
Encompass_Preferred

Biotin-PEG3-acid is a biotin-labeled PEG3 linker with a terminal carboxylic acid, supplied as a solid for use in conjugation and PROTAC linker synthesis. It provides a hydrophilic spacer arm to attach biotin to small molecules or probes, enabling affinity capture, purification, and detection workflows.

  • Biotin-labeled PEG3 linker with carboxylic acid terminus.
  • Suitable for coupling chemistries such as amide bond formation.
  • Used in PROTAC synthesis, biotinylation, pull-downs, and detection assays.
  • Molecular weight 447.55 g·mol⁻¹ and CAS 252881-76-8.
  • Purity 98.0%.
  • Supplied as a 250 MG solid; other sizes available.

Catalog No. 50-002-29481 Supplier Medchemexpress LLC Supplier No. HY116027250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG3-acid is a biotin-labeled PEG3 linker with a terminal carboxylic acid, supplied as a solid for use in conjugation and PROTAC linker synthesis. It provides a hydrophilic spacer arm to attach biotin to small molecules or probes, enabling affinity capture, purification, and detection workflows.

  • Biotin-labeled PEG3 linker with carboxylic acid terminus.
  • Suitable for coupling chemistries such as amide bond formation.
  • Used in PROTAC synthesis, biotinylation, pull-downs, and detection assays.
  • Molecular weight 447.55 g·mol⁻¹ and CAS 252881-76-8.
  • Purity 98.0%.
  • Supplied as a 250 MG solid; other sizes available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.