Learn More
Medchemexpress LLC Biotin-PEG3-acid 5g | 252881-76-8 | 447.55 | C19H33N3O7S | 5 G
SDP
Catalog No. 5000227226
Encompass_Preferred
Biotin-PEG3-acid is a biotin-labeled PEG linker with a terminal carboxylic acid group designed for use in bioconjugation and PROTAC synthesis. It provides a short, hydrophilic PEG3 spacer to improve solubility and reduce steric hindrance while enabling affinity capture or detection via the biotin tag.
  • Biotin-labeled PEG3 spacer for affinity capture and detection.
  • Terminal carboxylic acid enables straightforward amide coupling to amine-containing partners.
  • Hydrophilic PEG spacer improves solubility and linker flexibility.
  • Analytical data available (molecular weight 447.55, molecular formula C19H33N3O7S).
  • Supplied as a solid for research use, available in gram-scale quantities.
Catalog No. 50-002-27226 Supplier Medchemexpress LLC Supplier No. HY1160275G
Biotin-PEG3-acid is a biotin-labeled PEG linker with a terminal carboxylic acid group designed for use in bioconjugation and PROTAC synthesis. It provides a short, hydrophilic PEG3 spacer to improve solubility and reduce steric hindrance while enabling affinity capture or detection via the biotin tag.
  • Biotin-labeled PEG3 spacer for affinity capture and detection.
  • Terminal carboxylic acid enables straightforward amide coupling to amine-containing partners.
  • Hydrophilic PEG spacer improves solubility and linker flexibility.
  • Analytical data available (molecular weight 447.55, molecular formula C19H33N3O7S).
  • Supplied as a solid for research use, available in gram-scale quantities.