Learn More
Medchemexpress LLC Biotin-PEG3-aldehyde | 889443-90-7 | 95.0% | 550.67 g·mol⁻¹ | C26H38N4O7S | 50 MG
SDP
Catalog No. 5000203728
Encompass_Preferred
Biotin-PEG3-aldehyde is a cleavable three-unit polyethylene glycol (PEG) linker bearing a biotin tag and an aldehyde functional group. It is intended for use in antibody-drug conjugate (ADC) synthesis and bioconjugation where an aldehyde handle enables oxime or hydrazone bond formation.
  • Cleavable three-unit PEG linker with biotin tag.
  • Aldehyde functional group for oxime or hydrazone conjugation.
  • Available in multiple sizes, including 50 MG.
  • Typical purity around 95.0%.
  • Supplied as a solid powder; store powder at -20°C.
Catalog No. 50-002-03728 Supplier Medchemexpress LLC Supplier No. HY13605150MG
Biotin-PEG3-aldehyde is a cleavable three-unit polyethylene glycol (PEG) linker bearing a biotin tag and an aldehyde functional group. It is intended for use in antibody-drug conjugate (ADC) synthesis and bioconjugation where an aldehyde handle enables oxime or hydrazone bond formation.
  • Cleavable three-unit PEG linker with biotin tag.
  • Aldehyde functional group for oxime or hydrazone conjugation.
  • Available in multiple sizes, including 50 MG.
  • Typical purity around 95.0%.
  • Supplied as a solid powder; store powder at -20°C.