Learn More
Cayman Chemical BIotIn-PEG3-azIde 10mg
SDP
Catalog No. 502987859
Encompass_Preferred
A PEGylated version of biotin-azide that is more hydrophilic; can be used to label terminal alkynes via copper-catalyzed click reactions or copper-free click reactions with cyclooctynes; used to covalently tag proteins in combination with other tags
Catalog No. 50-298-7859 Supplier Cayman Chemical Supplier No. 2341910 MG
A PEGylated version of biotin-azide that is more hydrophilic; can be used to label terminal alkynes via copper-catalyzed click reactions or copper-free click reactions with cyclooctynes; used to covalently tag proteins in combination with other tags