Learn More
Selleck Chemical LLC Biotin-PEG3-azide E7219-5MG
SDP
Catalog No. 5000040981
Encompass_Preferred
Biotin-PEG3-azide is a PEG-based PROTAC linker can be used in the synthesis of PROTACs Biotin-PEG3-azide is a click chemistry reagent it contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-000-40981 Supplier Selleck Chemical LLC Supplier No. E72195MG
Biotin-PEG3-azide is a PEG-based PROTAC linker can be used in the synthesis of PROTACs Biotin-PEG3-azide is a click chemistry reagent it contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups