Learn More
Cayman Chemical Biotin-PEG3-azide, Purity: ≥95%, CAS: 875770-34-6, 50 mg
SDP
Catalog No. 502985754
Encompass_Preferred
A PEGylated version of biotin-azide that is more hydrophilic; can be used to label terminal alkynes via copper-catalyzed click reactions or copper-free click reactions with cyclooctynes; used to covalently tag proteins in combination with other tags
Catalog No. 50-298-5754 Supplier Cayman Chemical Supplier No. 2341950 MG
A PEGylated version of biotin-azide that is more hydrophilic; can be used to label terminal alkynes via copper-catalyzed click reactions or copper-free click reactions with cyclooctynes; used to covalently tag proteins in combination with other tags