Learn More
Medchemexpress LLC Biotin-PEG3-azide | 875770-34-6 | MFCD20134145 | ≥97.0% | 444.55 g/mol | C18H32N6O5S | 5 MG
SDP
Catalog No. 5000262213
Encompass_Preferred
Biotin-PEG3-azide is a PEGylated biotinylation reagent bearing a terminal azide group for copper-catalyzed click chemistry. It is employed as a linker in PROTAC synthesis and for affinity labeling and bioconjugation workflows requiring streptavidin capture.
  • Provides a three-unit PEG spacer for increased solubility and reduced steric hindrance.
  • Includes a terminal azide group compatible with click conjugation to alkynes.
  • Delivers a biotin affinity tag for efficient capture by streptavidin.
  • High purity suitable for research applications.
  • Soluble in DMSO and water with sonication as needed.
  • Stable as a powder when stored at low temperatures.
Catalog No. 50-002-62213 Supplier Medchemexpress LLC Supplier No. HY1301435MG
Biotin-PEG3-azide is a PEGylated biotinylation reagent bearing a terminal azide group for copper-catalyzed click chemistry. It is employed as a linker in PROTAC synthesis and for affinity labeling and bioconjugation workflows requiring streptavidin capture.
  • Provides a three-unit PEG spacer for increased solubility and reduced steric hindrance.
  • Includes a terminal azide group compatible with click conjugation to alkynes.
  • Delivers a biotin affinity tag for efficient capture by streptavidin.
  • High purity suitable for research applications.
  • Soluble in DMSO and water with sonication as needed.
  • Stable as a powder when stored at low temperatures.