Learn More
Medchemexpress LLC Biotin-PEG3-C3-NH2 | 183896-00-6 | 97.0% | C20H38N4O5S | 50 MG
SDP
Catalog No. 5000353166
Encompass_Preferred
Biotin-PEG3-C3-NH2 is a PEG-based PROTAC linker with an NH2 functional group, primarily used in the synthesis of PROTACs. This substance is intended for research use only, as a laboratory chemical for the manufacture of other substances.
  • PEG-based PROTAC linker with an NH2 functional group
  • Used in the synthesis of PROTACs
  • Displays fluorescence under streptavidin probes when combined with protein G
  • Can be used for high-throughput analysis of protein-small molecule interactions
  • Profiles enzyme activities in microarrays constructed by surface derivatization method
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-53166 Supplier Medchemexpress LLC Supplier No. HY140902A50MG
Biotin-PEG3-C3-NH2 is a PEG-based PROTAC linker with an NH2 functional group, primarily used in the synthesis of PROTACs. This substance is intended for research use only, as a laboratory chemical for the manufacture of other substances.
  • PEG-based PROTAC linker with an NH2 functional group
  • Used in the synthesis of PROTACs
  • Displays fluorescence under streptavidin probes when combined with protein G
  • Can be used for high-throughput analysis of protein-small molecule interactions
  • Profiles enzyme activities in microarrays constructed by surface derivatization method
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins