Learn More
Medchemexpress LLC Biotin-PEG3-CONH-Ph-CF3-diazirine, 10mg
SDP
Catalog No. NC3055977
Encompass_Preferred
Biotin-PEG3-CONH-Ph-CF3-diazirine is a biotin labeled PEG3 derivative. Biotin-PEG3-CONH-Ph-CF3-diazirine can be used for protein labeling.
Catalog No. NC3055977 Supplier Medchemexpress LLC Supplier No. HY15480110MG
Biotin-PEG3-CONH-Ph-CF3-diazirine is a biotin labeled PEG3 derivative. Biotin-PEG3-CONH-Ph-CF3-diazirine can be used for protein labeling.