Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-peg3-conh-ph-cf3-diazirine | 2845211-64-3 | C28H39F3N6O6S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1548015MG

Encompass_Preferred

Biotin-PEG3-CONH-Ph-CF3-diazirine is a biotin labeled PEG3 derivative that can be used for protein labeling. It has a high purity of 99.78% and a molecular weight of 644.71.

  • Biotin labeled PEG3 derivative.
  • Suitable for protein labeling.
  • High purity of 99.78%.
  • Molecular weight of 644.71.
  • Solid appearance, white to light yellow in color.
  • Soluble in DMSO at 100 mg/mL.
  • Stable for 3 years as powder at -20°C.
  • Stable for 6 months in solvent at -80°C.

Catalog No. 50-003-76902


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.