Learn More
Medchemexpress LLC Biotin-PEG3-iodoacetamide | 2306161-26-0 | 97.0% | 10 MG
SDP
Catalog No. 5000299129
Encompass_Preferred
Biotin-PEG3-iodoacetamide is a PEG-based linker combining a biotin tag, a three-unit PEG spacer, and an iodoacetamide reactive group. It selectively alkylates cysteine residues under mild conditions to introduce a biotin handle for affinity capture, detection, and enrichment workflows.
  • Selective cysteine labeling via iodoacetamide chemistry.
  • Biotin tag enables high-affinity streptavidin capture.
  • PEG3 spacer improves solubility and reduces steric hindrance.
  • Suitable for protein labeling, pull-downs, and detection assays.
  • Provided as a small-molecule reagent for research use.
Catalog No. 50-002-99129 Supplier Medchemexpress LLC Supplier No. HY17493810MG
Biotin-PEG3-iodoacetamide is a PEG-based linker combining a biotin tag, a three-unit PEG spacer, and an iodoacetamide reactive group. It selectively alkylates cysteine residues under mild conditions to introduce a biotin handle for affinity capture, detection, and enrichment workflows.
  • Selective cysteine labeling via iodoacetamide chemistry.
  • Biotin tag enables high-affinity streptavidin capture.
  • PEG3 spacer improves solubility and reduces steric hindrance.
  • Suitable for protein labeling, pull-downs, and detection assays.
  • Provided as a small-molecule reagent for research use.