Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG3-iodoacetamide | 2306161-26-0 | 97.0% | 586.48 | 100 MG
SDP

Catalog No. 5000384385
Encompass_Preferred

Biotin-PEG3-Iodoacetamide is a PEG derivative consisting of Biotin, 3 PEG units, and Iodoacetamide. Biotin forms a stable non-covalent bond with streptavidin, making this compound suitable for biochemical assays.

  • Composed of Biotin, 3 PEG units, and Iodoacetamide
  • Forms a stable non-covalent bond with streptavidin
  • Suitable for biochemical assay reagents
  • Soluble in DMSO: 100 mg/mL (ultrasonic)
  • Purity: 97.0%

Catalog No. 50-003-84385 Supplier Medchemexpress LLC Supplier No. 25MGHY174938
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG3-Iodoacetamide is a PEG derivative consisting of Biotin, 3 PEG units, and Iodoacetamide. Biotin forms a stable non-covalent bond with streptavidin, making this compound suitable for biochemical assays.

  • Composed of Biotin, 3 PEG units, and Iodoacetamide
  • Forms a stable non-covalent bond with streptavidin
  • Suitable for biochemical assay reagents
  • Soluble in DMSO: 100 mg/mL (ultrasonic)
  • Purity: 97.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.