Learn More
Medchemexpress LLC Biotin-PEG3-Mal | 1431618-70-0 | 98.9% | C25H39N5O8S | 50 MG
Supplier: Medchemexpress LLC HY14090750MG
Biotin-PEG3-Mal is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- Biotin-labeled
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- Exploits the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins