Learn More
Medchemexpress LLC Biotin-PEG3-NHS ester | 1253286-56-4 | 98.4% | 100 MG
SDP
Catalog No. 5000339140
Encompass_Preferred
Biotin-PEG3-NHS ester is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTAC (Proteolysis Targeting Chimeras) molecules. PROTACs typically consist of two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Serves as a linker in the construction of PROTACs
  • Linker is based on Polyethylene Glycol (PEG)
  • Intended for research purposes only
  • Primary application is in the synthesis of PROTACs
Catalog No. 50-003-39140 Supplier Medchemexpress LLC Supplier No. HY130291100MG
Biotin-PEG3-NHS ester is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTAC (Proteolysis Targeting Chimeras) molecules. PROTACs typically consist of two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Serves as a linker in the construction of PROTACs
  • Linker is based on Polyethylene Glycol (PEG)
  • Intended for research purposes only
  • Primary application is in the synthesis of PROTACs