Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG3-SS-azide | 2866429-93-6 | 99.6% | 692.91 | C27H48N8O7S3 | 10 MG
SDP

Catalog No. 5000203762
Encompass_Preferred

Biotin-PEG3-SS-azide is a cleavable three-unit polyethylene glycol linker bearing a biotin group and a terminal azide, designed for conjugation chemistry in research applications. It is suitable for click chemistry reactions and as a cleavable linker in the synthesis of antibody-drug conjugates.

  • Cleavable disulfide linker provides intracellular release.
  • Azide functionality enables CuAAC and strain-promoted cycloaddition.
  • PEG3 spacer increases solubility and reduces steric hindrance.
  • High purity supports reliable conjugation results.
  • Soluble in DMSO at high concentration for in vitro use.
  • Recommended storage at low temperature and protection from light to preserve stability.

Catalog No. 50-002-03762 Supplier Medchemexpress LLC Supplier No. HY14094410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG3-SS-azide is a cleavable three-unit polyethylene glycol linker bearing a biotin group and a terminal azide, designed for conjugation chemistry in research applications. It is suitable for click chemistry reactions and as a cleavable linker in the synthesis of antibody-drug conjugates.

  • Cleavable disulfide linker provides intracellular release.
  • Azide functionality enables CuAAC and strain-promoted cycloaddition.
  • PEG3 spacer increases solubility and reduces steric hindrance.
  • High purity supports reliable conjugation results.
  • Soluble in DMSO at high concentration for in vitro use.
  • Recommended storage at low temperature and protection from light to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.