Learn More
Medchemexpress LLC Biotin-PEG4-acid | 721431-18-1 | 97.0% | 1 G
SDP
Catalog No. 5000340809
Encompass_Preferred
Biotin-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to harness the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. They consist of two distinct ligands connected by a linker, with one ligand targeting an E3 ubiquitin ligase and the other targeting a protein of interest.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Facilitates selective degradation of target proteins
  • Engages the ubiquitin-proteasome system
Catalog No. 50-003-40809 Supplier Medchemexpress LLC Supplier No. HY1269591G
Biotin-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to harness the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. They consist of two distinct ligands connected by a linker, with one ligand targeting an E3 ubiquitin ligase and the other targeting a protein of interest.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Facilitates selective degradation of target proteins
  • Engages the ubiquitin-proteasome system