Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG4-acid | 721431-18-1 | 97.0% | 250 MG
SDP

Catalog No. 5000341788
Encompass_Preferred

Biotin-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This product is intended for research use only.

  • PEG-based PROTAC linker
  • Can be used in PROTAC synthesis
  • White to off-white solid appearance
  • Molecular weight: 491.60
  • Molecular formula: C21H37N3O8S
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-41788 Supplier Medchemexpress LLC Supplier No. 250MGHY126959
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This product is intended for research use only.

  • PEG-based PROTAC linker
  • Can be used in PROTAC synthesis
  • White to off-white solid appearance
  • Molecular weight: 491.60
  • Molecular formula: C21H37N3O8S
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.