Learn More
Medchemexpress LLC Biotin-PEG4-acid | 721431-18-1 | 97.0% | 5 G
SDP
Catalog No. 5000340883
Encompass_Preferred
Biotin-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTAC. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTAC
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40883 Supplier Medchemexpress LLC Supplier No. HY1269595G
Biotin-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTAC. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTAC
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins