Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-PEG4-acid | 721431-18-1 | 97.0% | 50 MG
SDP

Catalog No. 5000340825
Encompass_Preferred

Biotin-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound leverages the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins. It has a molecular weight of 491.60 and a chemical formula of C21H37N3O8S, appearing as a white to off-white solid.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • Molecular weight of 491.60
  • Chemical formula C21H37N3O8S
  • Appears as a white to off-white solid
  • Recommended for research use only

Catalog No. 50-003-40825 Supplier Medchemexpress LLC Supplier No. HY12695950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound leverages the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins. It has a molecular weight of 491.60 and a chemical formula of C21H37N3O8S, appearing as a white to off-white solid.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • Molecular weight of 491.60
  • Chemical formula C21H37N3O8S
  • Appears as a white to off-white solid
  • Recommended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.