Learn More
Medchemexpress LLC Biotin-PEG4-alkyne | 1458576-00-5 | 99.7% | C21H35N3O6S | 100 MG
SDP
Catalog No. 5000353175
Encompass_Preferred
Biotin-PEG4-alkyne is a biotin-labeled, PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. PROTACs consist of two ligands connected by a linker, facilitating selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Enables selective degradation of target proteins
Catalog No. 50-003-53175 Supplier Medchemexpress LLC Supplier No. HY140922100MG
Biotin-PEG4-alkyne is a biotin-labeled, PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. PROTACs consist of two ligands connected by a linker, facilitating selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • Biotin-labeled, PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Enables selective degradation of target proteins