Learn More
Medchemexpress LLC Biotin-PEG4-amide-Alkyne | 1006592-45-5 | 95.0% | C24H40N4O7S | 25 MG
SDP
Catalog No. 5000352684
Encompass_Preferred
Biotin-PEG4-amide-Alkyne is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Degrades target proteins via the ubiquitin-proteasome system
Catalog No. 50-003-52684 Supplier Medchemexpress LLC Supplier No. 25MGHY140923
Biotin-PEG4-amide-Alkyne is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Degrades target proteins via the ubiquitin-proteasome system