Learn More
Medchemexpress LLC Biotin-PEG4-amide-Alkyne | 1006592-45-5 | 96.41% | C24H40N4O7S | 50 MG
SDP
Catalog No. 5000354882
Encompass_Preferred
Biotin-PEG4-amide-Alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Alkyne group, which can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing azide groups
Catalog No. 50-003-54882 Supplier Medchemexpress LLC Supplier No. HY14092350MG
Biotin-PEG4-amide-Alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Alkyne group, which can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing azide groups