Learn More
Medchemexpress LLC Biotin-PEG4-Amide-C6-Azide | 1006592-62-6 | 99.1% | C27H49N7O7S | 100 MG
SDP
Catalog No. 5000353397
Encompass_Preferred
Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling specific reactions with other molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-53397 Supplier Medchemexpress LLC Supplier No. HY140914100MG
Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling specific reactions with other molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions