Learn More
Medchemexpress LLC Biotin-PEG4-Amide-C6-Azide | 1006592-62-6 | 99.1% | C27H49N7O7S | 25 MG
SDP
Catalog No. 5000352818
Encompass_Preferred
Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, facilitating copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne group molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN group molecules.
  • PEG-based PROTAC linker
  • Click chemistry reagent with azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-52818 Supplier Medchemexpress LLC Supplier No. 25MGHY140914
Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, facilitating copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne group molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN group molecules.
  • PEG-based PROTAC linker
  • Click chemistry reagent with azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)