Learn More
Medchemexpress LLC Biotin-PEG4-Amide-C6-Azide | 1006592-62-6 | 99.1% | C27H49N7O7S | 250 MG
SDP
Catalog No. 5000352871
Encompass_Preferred
Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups, or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52871 Supplier Medchemexpress LLC Supplier No. 250MGHY140914
Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups, or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins