Learn More
Medchemexpress LLC Biotin-PEG4-Amide-C6-Azide | 1006592-62-6 | 99.1% | C27H49N7O7S | 500 MG
SDP
Catalog No. 5000354879
Encompass_Preferred
Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-54879 Supplier Medchemexpress LLC Supplier No. HY140914500MG
Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups