Learn More
Medchemexpress LLC Biotin-PEG4-amide-C6-azide | 1006592-62-6 | MFCD30478293 | 99.1% | C27H49N7O7S | 10MG
SDP
Catalog No. 5000258608
Encompass_Preferred
Biotin-PEG4-amide-C6-azide is a biotinylated PEG4 amide linker terminating in an azide group, formulated as a high-purity research reagent for click chemistry and bioconjugation. It is suitable for linker assembly, affinity capture, and labeling workflows, and is soluble in DMSO and common co-solvent systems.
  • High purity suitable for analytical and preparative applications.
  • Terminal azide enables copper-catalyzed azide-alkyne cycloaddition for efficient conjugation.
  • PEG4 spacer provides flexibility and improved aqueous solubility.
  • Soluble in DMSO and compatible with typical in vivo co-solvent formulations.
  • Well suited for affinity capture, labeling, and linker assembly workflows.
Catalog No. 50-002-58608 Supplier Medchemexpress LLC Supplier No. HY14091410MG
Biotin-PEG4-amide-C6-azide is a biotinylated PEG4 amide linker terminating in an azide group, formulated as a high-purity research reagent for click chemistry and bioconjugation. It is suitable for linker assembly, affinity capture, and labeling workflows, and is soluble in DMSO and common co-solvent systems.
  • High purity suitable for analytical and preparative applications.
  • Terminal azide enables copper-catalyzed azide-alkyne cycloaddition for efficient conjugation.
  • PEG4 spacer provides flexibility and improved aqueous solubility.
  • Soluble in DMSO and compatible with typical in vivo co-solvent formulations.
  • Well suited for affinity capture, labeling, and linker assembly workflows.