Learn More
TARGETMOL CHEMICALS INC Biotin-PEG4-Amide-C6-Azide 2
SDP
Catalog No. 502878447
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker utilized in PROTAC synthesis. Purity 99.19%
Catalog No. 50-287-8447 Supplier TARGETMOL CHEMICALS INC Supplier No. T1758025MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Biotin-PEG4-Amide-C6-Azide is a PEG-based PROTAC linker utilized in PROTAC synthesis. Purity 99.19%