Learn More
Medchemexpress LLC Biotin-PEG4-amine | 663171-32-2 | 98.0% | C20H38N4O6S | 250 MG
SDP
Catalog No. 5000352851
Encompass_Preferred
Biotin-PEG4-amine is a biotin-labeled, PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. PROTACs consist of two different ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other to a target protein.
  • Biotin-labeled compound
  • PEG-based linker for PROTAC synthesis
  • Utilized in targeted protein degradation
  • Leverages the ubiquitin-proteasome system
  • Designed with two ligands for E3 ubiquitin ligase and target protein binding
Catalog No. 50-003-52851 Supplier Medchemexpress LLC Supplier No. 250MGHY140895
Biotin-PEG4-amine is a biotin-labeled, PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. PROTACs consist of two different ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other to a target protein.
  • Biotin-labeled compound
  • PEG-based linker for PROTAC synthesis
  • Utilized in targeted protein degradation
  • Leverages the ubiquitin-proteasome system
  • Designed with two ligands for E3 ubiquitin ligase and target protein binding