Learn More
Medchemexpress LLC Biotin-PEG4-azide | 1309649-57-7 | 99.8% | C20H36N6O6S | 25 MG
SDP
Catalog No. 5000352524
Encompass_Preferred
Biotin-PEG4-azide is a biotin-labeled, PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It functions as a versatile click chemistry reagent, facilitating both copper-catalyzed and strain-promoted cycloaddition reactions.
  • Biotin-labeled and PEG-based PROTAC linker
  • Essential for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-52524 Supplier Medchemexpress LLC Supplier No. 25MGHY140910
Biotin-PEG4-azide is a biotin-labeled, PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It functions as a versatile click chemistry reagent, facilitating both copper-catalyzed and strain-promoted cycloaddition reactions.
  • Biotin-labeled and PEG-based PROTAC linker
  • Essential for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups