Learn More
Medchemexpress LLC Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide | 2599839-59-3 | 95.0% | 978.24 | C47H67N9O10SSi | 5 MG
SDP
Catalog No. 5000221425
Encompass_Preferred
Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide is a clickable, acid-cleavable biotin-picolyl azide reagent designed for labeling and enrichment of cell-surface glycoproteins. It contains an azide functionality for bioorthogonal conjugation and is compatible with both copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), supplied as a solid for research applications.
  • Clickable azide functional group for CuAAC and SPAAC reactions.
  • Acid-cleavable dialkoxydiphenylsilane linker enables controlled release after enrichment.
  • Biotin tag enables affinity enrichment using streptavidin or avidin systems.
  • PEG4 spacer improves solubility and reduces steric hindrance during conjugation.
  • High purity (95.0%) for consistent labeling performance.
  • Suitable for proteomics workflows targeting cell-surface glycoproteins.
Catalog No. 50-002-21425 Supplier Medchemexpress LLC Supplier No. HY1453835MG
Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide is a clickable, acid-cleavable biotin-picolyl azide reagent designed for labeling and enrichment of cell-surface glycoproteins. It contains an azide functionality for bioorthogonal conjugation and is compatible with both copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), supplied as a solid for research applications.
  • Clickable azide functional group for CuAAC and SPAAC reactions.
  • Acid-cleavable dialkoxydiphenylsilane linker enables controlled release after enrichment.
  • Biotin tag enables affinity enrichment using streptavidin or avidin systems.
  • PEG4 spacer improves solubility and reduces steric hindrance during conjugation.
  • High purity (95.0%) for consistent labeling performance.
  • Suitable for proteomics workflows targeting cell-surface glycoproteins.