Learn More
Medchemexpress LLC Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide | 2599839-59-3 | 95.0% | C47H67N9O10SSi | 25 MG
SDP
Catalog No. 5000356608
Encompass_Preferred
Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide is a clickable, acid-cleavable biotin-picolyl azide. It functions as an enrichment handle for cell surface glycoproteins, facilitating protein labeling.
  • Clickable, acid-cleavable biotin-picolyl azide
  • Functions as an enrichment handle for cell surface glycoproteins
  • Suitable for protein labeling
  • Azide group enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-56608 Supplier Medchemexpress LLC Supplier No. 25MGHY145383
Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide is a clickable, acid-cleavable biotin-picolyl azide. It functions as an enrichment handle for cell surface glycoproteins, facilitating protein labeling.
  • Clickable, acid-cleavable biotin-picolyl azide
  • Functions as an enrichment handle for cell surface glycoproteins
  • Suitable for protein labeling
  • Azide group enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC)