Learn More
Medchemexpress LLC Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide | 2599839-59-3 | 95.0% | C47H67N9O10SSi | 50 MG
SDP
Catalog No. 5000354205
Encompass_Preferred
Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide is a clickable and acid-cleavable biotin-picolyl azide. It functions as an enrichment handle for cell surface glycoproteins, facilitating protein labeling by enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Clickable and acid-cleavable biotin-picolyl azide
  • Functions as an enrichment handle for cell surface glycoproteins
  • Facilitates protein labeling
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-54205 Supplier Medchemexpress LLC Supplier No. HY14538350MG
Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide is a clickable and acid-cleavable biotin-picolyl azide. It functions as an enrichment handle for cell surface glycoproteins, facilitating protein labeling by enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Clickable and acid-cleavable biotin-picolyl azide
  • Functions as an enrichment handle for cell surface glycoproteins
  • Facilitates protein labeling
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups