Learn More
Medchemexpress LLC Biotin-PEG4-dialkoxydiphenylsilane-picolyl azide | 2599839-59-3 | MFCD00137487 | 95.0% | C47H67N9O10SSi | 10MG
SDP
Catalog No. 5000260978
Encompass_Preferred
A clickable, acid-cleavable biotinylated PEG4 azide designed as an enrichment handle for labeling and enriching cell-surface glycoproteins. It incorporates a picolyl azide for efficient bioorthogonal conjugation and a dialkoxydiphenylsilane linker that permits acid-triggered release following affinity capture, providing a workflow for selective labeling, capture, and release of target biomolecules.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for covalent labeling.
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free reactions.
  • Acid-cleavable dialkoxydiphenylsilane linker allows release after enrichment.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Biotin enables strong affinity capture with streptavidin reagents.
  • Supplied with typical purity of 95% for consistent labeling results.
Catalog No. 50-002-60978 Supplier Medchemexpress LLC Supplier No. HY14538310MG
A clickable, acid-cleavable biotinylated PEG4 azide designed as an enrichment handle for labeling and enriching cell-surface glycoproteins. It incorporates a picolyl azide for efficient bioorthogonal conjugation and a dialkoxydiphenylsilane linker that permits acid-triggered release following affinity capture, providing a workflow for selective labeling, capture, and release of target biomolecules.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for covalent labeling.
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free reactions.
  • Acid-cleavable dialkoxydiphenylsilane linker allows release after enrichment.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Biotin enables strong affinity capture with streptavidin reagents.
  • Supplied with typical purity of 95% for consistent labeling results.