Learn More
Medchemexpress LLC Biotin-PEG4-hydrazide | 756525-97-0 | 99.9% | C21H39N5O7S | 100 MG
SDP
Catalog No. 5000355074
Encompass_Preferred
Biotin-PEG4-hydrazide is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Enables selective degradation of target proteins via the intracellular ubiquitin-proteasome system
Catalog No. 50-003-55074 Supplier Medchemexpress LLC Supplier No. HY140932100MG
Biotin-PEG4-hydrazide is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Enables selective degradation of target proteins via the intracellular ubiquitin-proteasome system